ENA336955829 Enamine

5-methyl-2-[methyl(propan-2-yl)amino]benzoic acid

  • MDL number MFCD16783242
  • Cas Number : 1247469-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84360
Availability details for
Quantity
  • Availability
  • Add to Cart