ENA336955832 Enamine

5-bromo-N-(2-hydroxyethyl)pyridine-2-carboxamide

  • MDL number MFCD17259298
  • Cas Number : 1289059-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84379
Availability details for
Quantity
  • Availability
  • Add to Cart