ENA336955841 Enamine

1,3-difluoro-5-methoxy-2-nitrobenzene

  • MDL number MFCD09029981
  • Cas Number : 66684-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84396
Availability details for
Quantity
  • Availability
  • Add to Cart