ENA336955849 Enamine

3-[(tert-butylcarbamoyl)amino]benzoic acid

  • MDL number MFCD11171982
  • Cas Number : 501679-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84410

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84410
Availability details for
Quantity
  • Availability
  • Add to Cart