ENA336955850 Enamine

1-[2-(morpholin-4-yl)phenyl]ethan-1-amine

  • MDL number MFCD11137043
  • Cas Number : 874993-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84412
Availability details for
Quantity
  • Availability
  • Add to Cart