ENA336955854 Enamine

4-[(2,5-dioxopyrrolidin-3-yl)methyl]benzoic acid

  • MDL number MFCD20233488
  • Cas Number : 1333753-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84473
Availability details for
Quantity
  • Availability
  • Add to Cart