ENA336955858 Enamine

5-[(2,2-dimethylpropyl)carbamoyl]pyridine-2-carboxylic acid

  • MDL number MFCD13216857
  • Cas Number : 1307267-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84480
Availability details for
Quantity
  • Availability
  • Add to Cart