ENA336955863 Enamine

1-(1-methyl-1H-pyrazole-4-sulfonyl)piperidin-4-amine hydrochloride

  • MDL number MFCD20233493
  • Cas Number : 1333799-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237...239
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84490
Availability details for
Quantity
  • Availability
  • Add to Cart