ENA336955864 Enamine

3-[3-(1-aminoethyl)phenyl]-1,3-oxazolidin-2-one

  • MDL number MFCD13196347
  • Cas Number : 1210238-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56858

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56858
Availability details for
Quantity
  • Availability
  • Add to Cart