ENA336955866 Enamine

4-[(propan-2-yl)sulfamoyl]-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09053259
  • Cas Number : 937613-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84494
Availability details for
Quantity
  • Availability
  • Add to Cart