ENA336955874 Enamine

2-(propan-2-yl)-1H-imidazole-4-sulfonyl chloride

  • MDL number MFCD19663587
  • Cas Number : 1334000-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84502
Availability details for
Quantity
  • Availability
  • Add to Cart