ENA336955876 Enamine

(2,2-dimethylpropyl)(methyl)amine

  • MDL number MFCD00048257
  • Cas Number : 26153-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57315

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-57315
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart