ENA336955880 Enamine

N-[2-(pyridin-2-yl)ethyl]cyclopropanamine

  • MDL number MFCD12797478
  • Cas Number : 1182884-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84508
Availability details for
Quantity
  • Availability
  • Add to Cart