ENA336955882 Enamine

4-methyl-3-[(oxan-4-yl)methoxy]aniline

  • MDL number MFCD16754992
  • Cas Number : 1247243-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84514
Availability details for
Quantity
  • Availability
  • Add to Cart