ENA336955884 Enamine

2-chlorocyclohex-1-ene-1-carbonitrile

  • MDL number MFCD11502628
  • Cas Number : 119205-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38...40
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84516

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84516
Availability details for
Quantity
  • Availability
  • Add to Cart