ENA336955886 Enamine

2-(4-methylphenyl)piperidine

  • MDL number MFCD02663600
  • Cas Number : 85237-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84530
Availability details for
Quantity
  • Availability
  • Add to Cart