ENA336955903 Enamine

2-(4-fluorophenyl)cyclopentan-1-one

  • MDL number MFCD15527061
  • Cas Number : 1014718-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84560

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84560
Availability details for
Quantity
  • Availability
  • Add to Cart