ENA336955909 Enamine

3-fluoro-4-sulfanylbenzonitrile

  • MDL number MFCD12109900
  • Cas Number : 110888-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84582
Availability details for
Quantity
  • Availability
  • Add to Cart