ENA336955910 Enamine

N-benzyl-N-cyclopropylpiperidine-4-carboxamide

  • MDL number MFCD12533996
  • Cas Number : 1154254-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84583

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84583
Availability details for
Quantity
  • Availability
  • Add to Cart