ENA336955912 Enamine

{[6-(difluoromethoxy)-1,3-dioxaindan-5-yl]methyl}(methyl)amine

  • MDL number MFCD16061332
  • Cas Number : 1249877-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84585
Availability details for
Quantity
  • Availability
  • Add to Cart