ENA336955915 Enamine

1-amino-4-methylpentan-3-ol

  • MDL number MFCD16300694
  • Cas Number : 860546-11-8
  • Molecular Weight : 117.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-84592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84592
Availability details for
Quantity
  • Availability
  • Add to Cart