ENA336955917 Enamine

2-amino-1-(1-methyl-1H-pyrrol-2-yl)ethan-1-ol

  • MDL number MFCD09041281
  • Cas Number : 926248-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84622
Availability details for
Quantity
  • Availability
  • Add to Cart