ENA336955923 Enamine

3-(5-amino-3-cyclopropyl-1H-pyrazol-1-yl)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD11209800
  • Cas Number : 1152665-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84630
Availability details for
Quantity
  • Availability
  • Add to Cart