ENA336955932 Enamine

methyl 3-amino-3-(2,4-difluorophenyl)propanoate hydrochloride

  • MDL number MFCD20233505
  • Cas Number : 1333750-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84657

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84657
Availability details for
Quantity
  • Availability
  • Add to Cart