ENA336955933 Enamine

3-bromo-1-phenyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD11983355
  • Cas Number : 72000-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84659
Availability details for
Quantity
  • Availability
  • Add to Cart