ENA336955936 Enamine

3-amino-3-(2,4-difluorophenyl)propanoic acid

  • MDL number MFCD02663205
  • Cas Number : 412925-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84663

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84663
Availability details for
Quantity
  • Availability
  • Add to Cart