ENA336955943 Enamine

1-(2-methanesulfonylethyl)-1H-pyrazol-3-amine

  • MDL number MFCD12806107
  • Cas Number : 1093967-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84672
Availability details for
Quantity
  • Availability
  • Add to Cart