ENA336955947 Enamine

4-cyano-2-fluorobenzene-1-sulfonamide

  • MDL number MFCD16093753
  • Cas Number : 918967-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84683
Availability details for
Quantity
  • Availability
  • Add to Cart