ENA336955948 Enamine

1-(4-tert-butylphenyl)-5-propyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD13654566
  • Cas Number : 1096911-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84688

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84688
Availability details for
Quantity
  • Availability
  • Add to Cart