ENA336955957 Enamine

tert-butyl 2,5-dimethylpiperazine-1-carboxylate

  • MDL number MFCD12761575
  • Cas Number : 643041-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84712

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84712
Availability details for
Quantity
  • Availability
  • Add to Cart