ENA336955970 Enamine

2-fluoro-N-(piperidin-4-yl)-N-(2,2,2-trifluoroethyl)benzamide

  • MDL number MFCD13737727
  • Cas Number : 1155143-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84737
Availability details for
Quantity
  • Availability
  • Add to Cart