ENA336955971 Enamine

1-[3-(propan-2-yl)phenyl]-5-propyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD13654555
  • Cas Number : 1097050-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84738
Availability details for
Quantity
  • Availability
  • Add to Cart