ENA336955979 Enamine

tert-butyl N-(carbamothioylmethyl)-N-ethylcarbamate

  • MDL number MFCD16829988
  • Cas Number : 1249334-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64272
Availability details for
Quantity
  • Availability
  • Add to Cart