ENA336955980 Enamine

1-methyl-1H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD10696345
  • Cas Number : 16681-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65210

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65210
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart