ENA336955983 Enamine

N-(piperidin-3-yl)benzamide

  • MDL number MFCD09805381
  • Cas Number : 55306-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66294
Availability details for
Quantity
  • Availability
  • Add to Cart