ENA336955987 Enamine

N,3-dihydroxy-4-methylbenzamide

  • MDL number MFCD09938400
  • Cas Number : 1016847-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84768

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84768
Availability details for
Quantity
  • Availability
  • Add to Cart