ENA336955995 Enamine

1-(5-methylpyridin-2-yl)piperidin-4-amine

  • MDL number MFCD09732472
  • Cas Number : 596817-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84779
Availability details for
Quantity
  • Availability
  • Add to Cart