ENA336956010 Enamine

5-(1-chloroethyl)-3-(2-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD13358693
  • Cas Number : 1040053-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25936
Availability details for
Quantity
  • Availability
  • Add to Cart