ENA336956016 Enamine

6-methylimidazo[2,1-b][1,3]thiazole-5-carbonitrile

  • MDL number MFCD12403317
  • Cas Number : 83253-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84823

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84823
Availability details for
Quantity
  • Availability
  • Add to Cart