ENA336956017 Enamine

4-chloro-8-methylquinoline-3-carbonitrile

  • MDL number MFCD09261373
  • Cas Number : 936497-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84824
Availability details for
Quantity
  • Availability
  • Add to Cart