ENA336956030 Enamine

2-[(2-bromoethoxy)methyl]oxane

  • MDL number MFCD09814347
  • Cas Number : 1016507-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84850
Availability details for
Quantity
  • Availability
  • Add to Cart