ENA336956031 Enamine

1-ethyl-N,3-dimethylpiperidin-4-amine

  • MDL number MFCD14692733
  • Cas Number : 1247239-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84851
Availability details for
Quantity
  • Availability
  • Add to Cart