ENA336956035 Enamine

1-[(5-ethyl-1,2,4-oxadiazol-3-yl)methyl]-N-methylpiperidin-3-amine

  • MDL number MFCD16789026
  • Cas Number : 1249225-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71814
Availability details for
Quantity
  • Availability
  • Add to Cart