ENA336956036 Enamine

2-amino-5H,6H,7H-cyclopenta[b]pyridine-3-carboxylic acid

  • MDL number MFCD16852616
  • Cas Number : 115271-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84860
Availability details for
Quantity
  • Availability
  • Add to Cart