ENA336956053 Enamine

2,6-dichlorobenzene-1-sulfonamide

  • MDL number MFCD03093819
  • Cas Number : 10290-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26994

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26994
Availability details for
Quantity
  • Availability
  • Add to Cart