ENA336956055 Enamine

1-(2-bromoethoxy)-3,5-dichlorobenzene

  • MDL number MFCD12093630
  • Cas Number : 18800-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84892
Availability details for
Quantity
  • Availability
  • Add to Cart