ENA336956059 Enamine

3-(4-methylphenyl)pent-2-enoic acid

  • Cas Number : 174660-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84897
Availability details for
Quantity
  • Availability
  • Add to Cart