ENA336956060 Enamine

2-(octyloxy)acetic acid

  • MDL number MFCD11196116
  • Cas Number : 63632-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84898
Availability details for
Quantity
  • Availability
  • Add to Cart