ENA336956065 Enamine

3,4-dimethyl pyridine-3,4-dicarboxylate

  • MDL number MFCD00160293
  • Cas Number : 1796-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-165
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84950
Availability details for
Quantity
  • Availability
  • Add to Cart