ENA336956068 Enamine

2-{[(4-oxo-3,4-dihydroquinazolin-2-yl)methyl]sulfanyl}propanoic acid

  • MDL number MFCD11540681
  • Cas Number : 1097090-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84954
Availability details for
Quantity
  • Availability
  • Add to Cart